1,5-Dibromo-2-iodo-3,4-dimethylbenzene ≥95%

Supplier: AOBChem USA

Warning

45424-500MG 45424-1G 45424-5G 45424-25G
77326-924EA 183.34 USD
77326-924 77347-116 77363-214 77385-928
1,5-Dibromo-2-iodo-3,4-dimethylbenzene ≥95%
1,5-Dibromo-2-iodo-3,4-dimethylbenzene

Precaución: for Research Use Only

Formula: C₈H₇Br₂I
MW: 389.854 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846012
CAS Number: 1160573-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR