N-(4-Bromo-3-methoxyphenyl)methanesulfonamide ≥97%

Supplier: AOBChem USA

Warning

45418-500MG 45418-1G 45418-5G 45418-25G
77326-922EA 182.31 USD
77326-922 77346-948 77363-210 77385-926
N-(4-Bromo-3-methoxyphenyl)methanesulfonamide ≥97%
N-(4-Bromo-3-methoxyphenyl)methanesulfonamide

Precaución: for Research Use Only

Formula: C₈H₁₀BrNO₃S
MW: 280.139 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23729302
CAS Number: 177360-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR