1-(Benzyloxy)-4-bromo-5-fluoro-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

45390-500MG 45390-1G 45390-25G 45390-5G
77326-920EA 304.88 USD
77326-920 77350-552 77385-924 77374-754
1-(Benzyloxy)-4-bromo-5-fluoro-2-methylbenzene ≥97%
1-(Benzyloxy)-4-bromo-5-fluoro-2-methylbenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrFO
MW: 295.147 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902673
CAS Number: 2234291-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR