4-Chloro-5-fluoro-2-methylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

44890-500MG 44890-1G 44890-5G
77339-306EA 575.77 USD
77339-306 77350-548 77374-750
4-Chloro-5-fluoro-2-methylphenylboronic acid pinacol ester ≥97%
4-Chloro-5-fluoro-2-methylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BClFO₂
MW: 270.535 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24039589
CAS Number: 1885096-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR