4-Carboxy-2,5-difluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

44778-500MG 44778-1G 44778-5G
77337-126EA 408.91 USD
77337-126 77350-546 77374-748
4-Carboxy-2,5-difluorophenylboronic acid ≥97%
4-Carboxy-2,5-difluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BF₂O₄
MW: 201.92 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28515386
CAS Number: 1966890-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR