(2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

Synonyms: [2-Chloro-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanol

43614-500MG 43614-1G 43614-5G 43614-25G
77326-914EA 161.71 USD
77326-914 77345-872 77363-204 77385-920
(2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol ≥95%
(2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₃H₁₈BClO₃
MW: 268.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18729958
CAS Number: 1799974-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR