(3-Chloro-4-methanesulfonylphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

42667-1G 42667-5G 42667-25G
77341-458EA 87.55 USD
77341-458 77363-198 77374-738
(3-Chloro-4-methanesulfonylphenyl)boronic acid ≥97%
(3-Chloro-4-methanesulfonylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₈BClO₄S
MW: 234.465 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27935715
CAS Number: 2239305-86-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR