(4-Methanesulfonyl-3-methoxyphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

42664-500MG 42664-1G 42664-5G
77326-906EA 313.12 USD
77326-906 77350-536 77374-736
(4-Methanesulfonyl-3-methoxyphenyl)boronic acid ≥97%
(4-Methanesulfonyl-3-methoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₅S
MW: 230.046 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27935712
CAS Number: 1313617-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR