(6-Chloro-2H-1,3-benzodioxol-5-yl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

42529-500MG 42529-1G 42529-5G
77326-904EA 318.27 USD
77326-904 77350-534 77374-734
(6-Chloro-2H-1,3-benzodioxol-5-yl)boronic acid ≥95%
(6-Chloro-2H-1,3-benzodioxol-5-yl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₆BClO₄
MW: 200.384 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27935571
CAS Number: 2377608-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR