3-Chloro-4-(cyclopropylmethoxy)phenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

42383-500MG 42383-1G 42383-5G 42383-25G
77326-902EA 88.58 USD
77326-902 77342-324 77363-196 77374-732
3-Chloro-4-(cyclopropylmethoxy)phenylboronic acid ≥95%
3-Chloro-4-(cyclopropylmethoxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂BClO₃
MW: 226.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18383681
CAS Number: 1228181-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR