1,2,3-trichloro-4-(methylsulfonyl)benzene ≥97%

Supplier: AOBChem USA

Warning

42316-1G 42316-5G 42316-25G
77341-306EA 82.4 USD
77341-306 77363-194 77374-730
1,2,3-trichloro-4-(methylsulfonyl)benzene ≥97%
1,2,3-trichloro-4-(methylsulfonyl)benzene

Precaución: for Research Use Only

Formula: C₇H₅Cl₃O₂S
Storage Temperature: Ambient
MDL Number: MFCD34756211
CAS Number: 176375-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR