2-Cyano-3-methoxybenzeneboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-Cyano-3-methoxyphenylboronic acid

42039-500MG 42039-1G 42039-5G
77326-898EA 335.78 USD
77326-898 77350-530 77374-728
2-Cyano-3-methoxybenzeneboronic acid ≥95%
2-Cyano-3-methoxybenzeneboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BNO₃
Storage Temperature: Ambient
MDL Number: MFCD10696671
CAS Number: 1164100-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR