2-Cyano-4-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

41973-500MG 41973-1G 41973-5G 41973-25G
77326-896EA 263.68 USD
77326-896 77350-528 77363-190 77385-910
2-Cyano-4-methoxybenzoic acid ≥97%
2-Cyano-4-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₇NO₃
Storage Temperature: Ambient
CAS Number: 22246-20-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR