2-Chloro-4-nitrophenylboronic acid, pinacol ester ≥97%

Supplier: AOBChem USA

Warning

41125-500MG 41125-1G 41125-5G 41125-25G
77326-890EA 196.73 USD
77326-890 77350-526 77363-182 77385-908
2-Chloro-4-nitrophenylboronic acid, pinacol ester ≥97%
2-Chloro-4-nitrophenylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₅BClNO₄
MW: 283.516 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20529521
CAS Number: 1426804-80-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR