4-Bromo-2-ethoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

40940-500MG 40940-1G 40940-5G 40940-25G
77326-886EA 187.46 USD
77326-886 77350-522 77363-178 77385-904
4-Bromo-2-ethoxyphenylboronic acid ≥97%
4-Bromo-2-ethoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BBrO₃
MW: 244.878 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21609537
CAS Number: 2096329-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR