4-Fluoro-3-methoxy-5-(trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

40906-1G 40906-5G 40906-25G
77341-866EA 106.09 USD
77341-866 77363-174 77374-718
4-Fluoro-3-methoxy-5-(trifluoromethyl)phenylboronic acid ≥97%
4-Fluoro-3-methoxy-5-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₇BF₄O₃
Storage Temperature: Ambient
MDL Number: MFCD21332932
CAS Number: 1701449-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR