5-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-(Bromomethyl)-2-fluorophenylboronic acid, pinacol ester, 2-(5-(Bromomethyl)-2-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

39995-1G 39995-5G 39995-25G
77340-670EA 60.77 USD
77340-670 77363-164 77374-706
5-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester ≥97%
5-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BBrFO₂
Storage Temperature: Ambient
MDL Number: MFCD17926519
CAS Number: 1256360-47-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR