4'-Bromo-2-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

39982-500MG 39982-1G 39982-5G 39982-25G
77326-878EA 161.71 USD
77326-878 77345-870 77363-162 77385-900
4'-Bromo-2-methyl-1,1'-biphenyl ≥97%
4'-Bromo-2-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₁Br
MW: 247.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26632020
CAS Number: 106475-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR