4-Chloro-2-fluoro-5-hydroxyphenylboronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

39974-500MG 39974-1G 39974-5G
77326-876EA 345.05 USD
77326-876 77350-518 77374-704
4-Chloro-2-fluoro-5-hydroxyphenylboronic acid pinacol ester ≥95%
4-Chloro-2-fluoro-5-hydroxyphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₅BClFO₃
MW: 272.508 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16295125
CAS Number: 1256360-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR