2-Chloro-5-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
2-Chloro-5-methylpyridine-3-boronic acid pinacol ester
39196-500MG
39196-1G
39196-5G
39196-25G
77326-872EA
195.7
USD
77326-872
77350-516
77363-150
77385-896
2-Chloro-5-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥97%
2-Chloro-5-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Precaución: for Research Use Only
Formula:
C₁₂H₁₇BClNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD08063083 CAS Number: 1309982-62-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...