5-Formyl-4-methylthiophene-2-carboxylic acid ≥95%
Supplier: AOBChem USA
|
Warning
|
39126-500MG
39126-1G
39126-5G
77336-216EA
383.16
USD
77336-216
77350-514
77374-694
5-Formyl-4-methylthiophene-2-carboxylic acid ≥95%
5-Formyl-4-methylthiophene-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₇H₆O₃S MW: 170.186 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD15730654 CAS Number: 391936-74-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...