1-Bromo-2-ethoxy-4-fluoro-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

38841-500MG 38841-1G 38841-5G 38841-25G
77326-870EA 141.11 USD
77326-870 77344-802 77363-146 77385-894
1-Bromo-2-ethoxy-4-fluoro-5-methylbenzene ≥97%
1-Bromo-2-ethoxy-4-fluoro-5-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrFO
MW: 233.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756194
CAS Number: 2387062-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR