4-Chloro-3-fluoro-2-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

38783-500MG 38783-1G 38783-5G
77337-074EA 407.88 USD
77337-074 77350-510 77374-690
4-Chloro-3-fluoro-2-methoxyphenol ≥97%
4-Chloro-3-fluoro-2-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₆ClFO₂
MW: 176.573 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22415088
CAS Number: 1783512-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR