(4-Chloro-3-fluoro-2-methoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

38772-500MG 38772-1G 38772-5G
77340-000EA 710.7 USD
77340-000 77350-508 77374-686
(4-Chloro-3-fluoro-2-methoxyphenyl)methanol ≥97%
(4-Chloro-3-fluoro-2-methoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₈ClFO₂
MW: 190.599 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28523090
CAS Number: 1780616-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR