1-Bromo-3-isopropoxy-5-isopropylbenzene ≥97%

Supplier: AOBChem USA

Warning

38768-500MG 38768-1G 38768-5G 38768-25G
77326-864EA 187.46 USD
77326-864 77350-506 77363-138 77385-888
1-Bromo-3-isopropoxy-5-isopropylbenzene ≥97%
1-Bromo-3-isopropoxy-5-isopropylbenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₇BrO
MW: 257.167 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28356553
CAS Number: 1369811-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR