(3-Bromo-5-isopropyl-2-methoxyphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

38759-500MG 38759-1G 38759-5G 38759-25G
77326-862EA 204.97 USD
77326-862 77350-502 77363-136 77385-886
(3-Bromo-5-isopropyl-2-methoxyphenyl)(methyl)sulfane ≥95%
(3-Bromo-5-isopropyl-2-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrOS
Storage Temperature: Ambient
MDL Number: MFCD34529953
CAS Number: 2734776-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR