Ethyl 2,6-dichloro-4-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

38755-500MG 38755-1G 38755-5G
77335-836EA 367.71 USD
77335-836 77350-500 77374-682
Ethyl 2,6-dichloro-4-fluorobenzoate ≥97%
Ethyl 2,6-dichloro-4-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₇Cl₂FO₂
MW: 237.055 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28346556
CAS Number: 1803820-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR