3-Bromo-5-ethyl-2-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

38750-500MG 38750-1G 38750-5G 38750-25G
77326-858EA 229.69 USD
77326-858 77350-498 77363-132 77385-882
3-Bromo-5-ethyl-2-hydroxybenzaldehyde ≥97%
3-Bromo-5-ethyl-2-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MW: 229.071 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06739516
CAS Number: 708209-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR