Ethyl 2,4-dichloro-5-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

38746-500MG 38746-1G 38746-5G
77326-856EA 333.72 USD
77326-856 77350-492 77374-676
Ethyl 2,4-dichloro-5-methylbenzoate ≥97%
Ethyl 2,4-dichloro-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀Cl₂O₂
MW: 233.091 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28786807
CAS Number: 1804896-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR