Ethyl 4-bromo-5-chloro-2-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

38742-500MG 38742-1G 38742-25G 38742-5G
77326-852EA 301.79 USD
77326-852 77350-488 77385-880 77374-672
Ethyl 4-bromo-5-chloro-2-methylbenzoate ≥97%
Ethyl 4-bromo-5-chloro-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrClO₂
Storage Temperature: Ambient
MDL Number: MFCD24387146
CAS Number: 1033436-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR