5-Chloro-3-ethoxy-2-fluoropyridine ≥97%

Supplier: AOBChem USA

Warning

38730-500MG 38730-1G 38730-5G 38730-25G
77326-850EA 116.39 USD
77326-850 77343-486 77363-128 77374-668
5-Chloro-3-ethoxy-2-fluoropyridine ≥97%
5-Chloro-3-ethoxy-2-fluoropyridine

Precaución: for Research Use Only

Formula: C₇H₇ClFNO
Storage Temperature: Ambient
MDL Number: MFCD32902662
CAS Number: 2586126-91-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR