4-Bromo-3-chloro-N-methoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

38727-500MG 38727-1G 38727-5G 38727-25G
77326-848EA 118.45 USD
77326-848 77343-554 77363-126 77374-666
4-Bromo-3-chloro-N-methoxy-N-methylbenzamide ≥97%
4-Bromo-3-chloro-N-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₉BrClNO₂
MW: 278.53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23133071
CAS Number: 1235711-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR