tert-Butyl 4-bromo-3-chlorobenzoate ≥97%

Supplier: AOBChem USA

Warning

38724-500MG 38724-1G 38724-5G 38724-25G
77326-846EA 204.97 USD
77326-846 77350-486 77363-124 77385-878
tert-Butyl 4-bromo-3-chlorobenzoate ≥97%
tert-Butyl 4-bromo-3-chlorobenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrClO₂
MW: 291.569 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23085724
CAS Number: 1494593-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR