Ethyl 3,6-difluoro-2-methoxybenzoate ≥95%

Supplier: AOBChem USA

Warning

38716-500MG 38716-1G 38716-5G
77326-844EA 325.48 USD
77326-844 77350-484 77374-664
Ethyl 3,6-difluoro-2-methoxybenzoate ≥95%
Ethyl 3,6-difluoro-2-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₂O₃
MW: 216.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18396278
CAS Number: 1261552-95-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR