4-Bromo-2,5-dimethylbenzonitrile ≥95%
Supplier: AOBChem USA
|
Warning
|
38710-500MG
38710-1G
38710-5G
38710-25G
77326-840EA
281.19
USD
77326-840
77350-482
77363-120
77385-874
4-Bromo-2,5-dimethylbenzonitrile ≥95%
4-Bromo-2,5-dimethylbenzonitrile
Precaución: for Research Use Only
Formula:
C₉H₈BrN MW: 210.071 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18905448 CAS Number: 292037-85-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...