5-Bromo-4-chloro-1-methyl-1H-indazole ≥95%

Supplier: AOBChem USA

Warning

38708-1G 38708-5G 38708-25G
77341-226EA 77.25 USD
77341-226 77363-118 77374-662
5-Bromo-4-chloro-1-methyl-1H-indazole ≥95%
5-Bromo-4-chloro-1-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₆BrClN₂
Storage Temperature: Ambient
MDL Number: MFCD33403791
CAS Number: 1785427-70-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR