(3-Bromo-2,5-dimethylphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

38702-500MG 38702-1G 38702-5G
77337-820EA 448.05 USD
77337-820 77350-476 77374-658
(3-Bromo-2,5-dimethylphenyl)methanol ≥95%
(3-Bromo-2,5-dimethylphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁BrO
Storage Temperature: Ambient
MDL Number: MFCD26403806
CAS Number: 247575-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR