4-Bromo-7-chloro-3-iodo-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

38693-500MG 38693-1G 38693-5G 38693-25G
77326-834EA 145.23 USD
77326-834 77345-070 77363-112 77385-870
4-Bromo-7-chloro-3-iodo-1H-indazole ≥97%
4-Bromo-7-chloro-3-iodo-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₃BrClIN₂
MW: 357.374 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30528346
CAS Number: 2385608-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR