Ethyl 4-chloro-3-(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

38691-500MG 38691-1G 38691-5G 38691-25G
77326-832EA 260.59 USD
77326-832 77350-474 77363-110 77385-868
Ethyl 4-chloro-3-(methylthio)benzoate ≥97%
Ethyl 4-chloro-3-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₂S
MW: 230.711 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22489167
CAS Number: 1822631-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR