Methyl 3-isopropoxy-4-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

38679-500MG 38679-1G 38679-5G 38679-25G
77326-826EA 157.59 USD
77326-826 77345-692 77363-106 77385-862
Methyl 3-isopropoxy-4-methylbenzoate ≥97%
Methyl 3-isopropoxy-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
MW: 208.254 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23115026
CAS Number: 865138-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR