Ethyl 2-bromo-3-chloro-6-fluorobenzoate ≥95%

Supplier: AOBChem USA

Warning

38668-500MG 38668-1G 38668-5G
77335-470EA 355.35 USD
77335-470 77350-466 77374-648
Ethyl 2-bromo-3-chloro-6-fluorobenzoate ≥95%
Ethyl 2-bromo-3-chloro-6-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₇BrClFO₂
Storage Temperature: Ambient
MDL Number: MFCD28141931
CAS Number: 1805478-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR