(2,6-Difluoro-3-iodophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

38665-500MG 38665-1G 38665-5G
77336-360EA 387.28 USD
77336-360 77350-464 77374-646
(2,6-Difluoro-3-iodophenyl)methanol ≥97%
(2,6-Difluoro-3-iodophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₅F₂IO
MW: 270.015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31635472
CAS Number: 1407118-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR