Methyl 2-fluoro-6-methoxy-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

38656-500MG 38656-1G 38656-5G
77336-358EA 387.28 USD
77336-358 77350-460 77374-640
Methyl 2-fluoro-6-methoxy-3-methylbenzoate ≥97%
Methyl 2-fluoro-6-methoxy-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃
MW: 198.191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707110
CAS Number: 2114517-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR