6-(Benzyloxy)-2-fluoro-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

38649-500MG 38649-1G 38649-5G 38649-25G
77326-814EA 152.44 USD
77326-814 77345-448 77363-094 77385-858
6-(Benzyloxy)-2-fluoro-3-methylbenzaldehyde ≥97%
6-(Benzyloxy)-2-fluoro-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₃FO₂
Storage Temperature: Ambient
MDL Number: MFCD32707109
CAS Number: 2432849-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR