Methyl 6-(benzyloxy)-2-fluoro-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

38648-500MG 38648-1G 38648-5G 38648-25G
77326-812EA 182.31 USD
77326-812 77346-946 77363-092 77385-856
Methyl 6-(benzyloxy)-2-fluoro-3-methylbenzoate ≥97%
Methyl 6-(benzyloxy)-2-fluoro-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₆H₁₅FO₃
MW: 274.287 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707108
CAS Number: 2484888-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR