4-Butoxy-2-fluoro-1-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

38638-500MG 38638-1G 38638-5G 38638-25G
77326-806EA 101.97 USD
77326-806 77342-988 77363-086 77374-630
4-Butoxy-2-fluoro-1-methylbenzene ≥97%
4-Butoxy-2-fluoro-1-methylbenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅FO
MW: 182.235 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707106
CAS Number: 1693661-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR