3-Acetyl-2,6-difluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

38633-1G 38633-5G 38633-25G
77340-954EA 64.89 USD
77340-954 77363-076 77374-628
3-Acetyl-2,6-difluorobenzoic acid ≥97%
3-Acetyl-2,6-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₆F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD26687990
CAS Number: 1503929-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR