6-Bromo-2-fluoro-3-isopropoxyphenol ≥97%

Supplier: AOBChem USA

Warning

38627-1G 38627-5G 38627-25G
77341-414EA 85.49 USD
77341-414 77363-074 77374-626
6-Bromo-2-fluoro-3-isopropoxyphenol ≥97%
6-Bromo-2-fluoro-3-isopropoxyphenol

Precaución: for Research Use Only

Formula: C₉H₁₀BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD32707104
CAS Number: 2413441-32-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR