Methyl 6-bromo-2-fluoro-3-isopropoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

38626-500MG 38626-1G 38626-5G 38626-25G
77326-796EA 243.08 USD
77326-796 77350-452 77363-072 77385-846
Methyl 6-bromo-2-fluoro-3-isopropoxybenzoate ≥97%
Methyl 6-bromo-2-fluoro-3-isopropoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrFO₃
MW: 291.114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902655
CAS Number: 2586125-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR