2-Fluoro-1-isopropoxy-4-(methoxymethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

38622-500MG 38622-1G 38622-5G
77336-060EA 375.95 USD
77336-060 77350-448 77374-622
2-Fluoro-1-isopropoxy-4-(methoxymethyl)benzene ≥97%
2-Fluoro-1-isopropoxy-4-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅FO₂
MW: 198.234 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34529947
CAS Number: 2734774-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR